accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3H3CCH3NCH3BOOH3CH3CCH3H3C

CAS RN 406726-92-9

IUPAC Name: 9-octyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Synonyms: 9-octyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Molecular Weight (g/mol): 531.35
Molecular Formula: C32H47B2NO4
InChi Key: XVVSDGVWSRLSDI-UHFFFAOYSA-N
SMILES: CCCCCCCCN1C2=C(C=CC(=C2)B2OC(C)(C)C(C)(C)O2)C2=C1C=C(C=C2)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results