accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOBrHO

CAS RN 4068-76-2

IUPAC Name: methyl 5-bromo-2-hydroxybenzoate
Synonyms: methyl 5-bromo-2-hydroxybenzoate
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: FJYDBKPPGRZSOZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results