accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOClHO

CAS RN 4068-78-4

IUPAC Name: methyl 5-chloro-2-hydroxybenzoate
Synonyms: methyl 5-chloro-2-hydroxybenzoate
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: KJWHRMZKJXOWFC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results