accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOHNOFFFCl

CAS RN 406930-04-9

IUPAC Name: 3-(2-chlorophenyl)-1-[4-(trifluoromethoxy)phenyl]urea
Synonyms: 3-(2-chlorophenyl)-1-[4-(trifluoromethoxy)phenyl]urea
Molecular Weight (g/mol): 330.69
Molecular Formula: C14H10ClF3N2O2
InChi Key: IBVFZYNARZAJSE-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(NC(=O)NC2=CC=CC=C2Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results