accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 407-01-2

IUPAC Name: bis(2,2,2-trifluoroethyl)amine
Synonyms: bis(2,2,2-trifluoroethyl)amine
Molecular Weight (g/mol): 181.08
Molecular Formula: C4H5F6N
InChi Key: GTJGHXLFPMOKCE-UHFFFAOYSA-N
SMILES: FC(F)(F)CNCC(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results