accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOOH2C

CAS RN 407-47-6

IUPAC Name: 2,2,2-trifluoroethyl prop-2-enoate
Synonyms: 2,2,2-trifluoroethyl prop-2-enoate
Molecular Weight (g/mol): 154.09
Molecular Formula: C5H5F3O2
InChi Key: VBHXIMACZBQHPX-UHFFFAOYSA-N
SMILES: FC(F)(F)COC(=O)C=C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)