accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3COOOO(E)

CAS RN 4070-80-8

IUPAC Name: sodium (2E)-4-(octadecyloxy)-4-oxobut-2-enoate
Synonyms: sodium stearyl fumarate
Molecular Weight (g/mol): 390.54
Molecular Formula: C22H39NaO4
InChi Key: STFSJTPVIIDAQX-LTRPLHCISA-M
SMILES: [Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C([O-])=O
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Grade 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)