accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOH

CAS RN 40731-98-4

IUPAC Name: 4-hydroxy-2,3-dihydro-1H-inden-1-one
Synonyms: 4-hydroxy-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: CKSCMRNFDBWFND-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1CCC2=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)