accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40733-28-6

IUPAC Name: 1-[(2R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[(2R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl]-5-methyl-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 356.49
Molecular Formula: C16H28N2O5Si
InChi Key: IJWIJLIIOKZJMS-YNEHKIRRSA-N
SMILES: CC1=CN([C@H]2C[C@H](O)[C@@H](CO[Si](C)(C)C(C)(C)C)O2)C(=O)NC1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results