accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4074-90-2

H2COOOOCH2

CAS RN 4074-90-2

IUPAC Name: 1,6-diethenyl hexanedioate
Synonyms: divinyl adipate
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: JZQAAQZDDMEFGZ-UHFFFAOYSA-N
SMILES: C=COC(=O)CCCCC(=O)OC=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results