accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40753-13-7

IUPAC Name: 4-(4-bromophenyl)-1,2,3-thiadiazole
Synonyms: 4-(4-bromophenyl)-1,2,3-thiadiazole
Molecular Weight (g/mol): 241.11
Molecular Formula: C8H5BrN2S
InChi Key: HGWOTVRPRHVJQK-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CSN=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results