accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOH

CAS RN 40771-26-4

IUPAC Name: 1,2,3,4-tetrahydronaphthalene-1,5-diol
Synonyms: 1,2,3,4-tetrahydronaphthalene-1,5-diol
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: MYIDTCFDQGAVFL-UHFFFAOYNA-N
SMILES: OC1CCCC2=C(O)C=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)