accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4080-98-2

IUPAC Name: dodecanedihydrazide
Synonyms: dodecanedihydrazide
Molecular Weight (g/mol): 258.37
Molecular Formula: C12H26N4O2
InChi Key: GRGBENNNGZARRZ-UHFFFAOYSA-N
SMILES: NNC(=O)CCCCCCCCCCC(=O)NN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results