accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONH

CAS RN 40807-61-2

IUPAC Name: 4-phenylpiperidin-4-ol
Synonyms: 4-phenylpiperidin-4-ol
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: KQKFQBTWXOGINC-UHFFFAOYSA-N
SMILES: OC1(CCNCC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)