accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3N(R)HOOH

CAS RN 40829-04-7

IUPAC Name: (2R)-1-hydroxy-3-(4-hydroxyphenyl)propan-2-aminium
Synonyms: (2R)-1-hydroxy-3-(4-hydroxyphenyl)propan-2-aminium
Molecular Weight (g/mol): 168.22
Molecular Formula: C9H14NO2
InChi Key: DBLDQZASZZMNSL-MRVPVSSYSA-O
SMILES: [NH3+][C@@H](CO)CC1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results