accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOO

CAS RN 40832-79-9

IUPAC Name: 1-(2-nitrophenyl)pyrrolidine
Synonyms: 1-(2-nitrophenyl)pyrrolidine
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: DHTSBEQPOXRZBB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results