accessibility menu, dialog, popup

UserName

Viewing profile as user:
HBrNH2NNH

CAS RN 40835-96-9

IUPAC Name: hydrogen 1H-perimidin-2-amine bromide
Synonyms: hydrogen 1H-perimidin-2-amine bromide
Molecular Weight (g/mol): 264.13
Molecular Formula: C11H10BrN3
InChi Key: HFZUTZWRTHBWPV-UHFFFAOYSA-N
SMILES: [H+].[Br-].NC1=NC2=CC=CC3=CC=CC(N1)=C23
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)