accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 408355-23-7

IUPAC Name: 7-bromo-5-fluoro-1H-indole
Synonyms: 7-bromo-5-fluoro-1H-indole
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H5BrFN
InChi Key: XOMFQRPHXMTJSG-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C2NC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results