accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4084-27-9

IUPAC Name: (4S,5R)-4,5,6-trihydroxy-2-oxohexanal
Synonyms: 3-deoxyglucosone 3-deoxy-D-fructose 3-deoxy-erythro-hexos-2-ulose 3-deoxy-2-ketoglucose 3-deoxy-D-erythro-hexosulose 3-deoxy-D-glucosone 3-deoxy-D-erythro-hexulose
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: ZGCHLOWZNKRZSN-NTSWFWBYSA-N
SMILES: OC[C@@H](O)[C@@H](O)CC(=O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results