accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CI

CAS RN 408492-28-4

IUPAC Name: 2-(3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 329.97
Molecular Formula: C12H16BIO2
InChi Key: HSHFNMSHDHAHDN-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(I)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results