accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOO

CAS RN 40870-59-5

IUPAC Name: (4-methyl-3-nitrophenyl)methanol
Synonyms: (4-methyl-3-nitrophenyl)methanol
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: URCWIFSXDARYLY-UHFFFAOYSA-N
SMILES: CC1=CC=C(CO)C=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)