accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH3

CAS RN 4088-22-6

IUPAC Name: methyldioctadecylamine
Synonyms: methyldioctadecylamine
Molecular Weight (g/mol): 536.03
Molecular Formula: C37H77N
InChi Key: VFLWKHBYVIUAMP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCN(C)CCCCCCCCCCCCCCCCCC
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results