accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)ONH2OHOCH3HCl

CAS RN 4089-07-0

IUPAC Name: ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride
Synonyms: L-tyrosine, ethyl ester hydrochloride
Molecular Weight (g/mol): 245.7
Molecular Formula: C11H16ClNO3
InChi Key: BQULAXAVRFIAHN-OXZMBGLCNA-N
SMILES: Cl.CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)