accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOCH3

CAS RN 4091-50-3

IUPAC Name: [2-(4-methoxyphenyl)ethyl](methyl)amine
Synonyms: [2-(4-methoxyphenyl)ethyl](methyl)amine
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: JCMWSVNNSPUNER-UHFFFAOYSA-N
SMILES: CNCCC1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results