accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3NH2HCl

CAS RN 40918-51-2

IUPAC Name: ethyl 2-aminopentanoate hydrochloride
Synonyms: ethyl 2-aminopentanoate hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: CHAJBHNQIZAJJM-UHFFFAOYNA-N
SMILES: Cl.CCCC(N)C(=O)OCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results