accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40925-68-6

IUPAC Name: 2-amino-4-bromophenol
Synonyms: 2-amino-4-bromophenol
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: JHRIPENGTGSNPJ-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CC=C1O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)