accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40932-63-6

IUPAC Name: 1-(2-methyl-5-phenylthiophen-3-yl)ethan-1-one
Synonyms: 1-(2-methyl-5-phenylthiophen-3-yl)ethanone
Molecular Weight (g/mol): 216.3
Molecular Formula: C13H12OS
InChi Key: GREAZYFTAJMZFD-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)SC(=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results