CAS RN 40953-34-2
IUPAC Name:
2-amino-1H-imidazole-4,5-dicarbonitrile
Synonyms:
2-amino-1H-imidazole-4,5-dicarbonitrile
Molecular Weight (g/mol):
133.11
Molecular Formula:
C5H3N5
InChi Key:
MLOXIXGLIZLPDP-UHFFFAOYSA-N
SMILES:
NC1=NC(C#N)=C(N1)C#N
1
–
1
of
1
results
1
2-Amino-4,5-dicyano-1H-imidazole 97.0+%, TCI America™
CAS: 40953-34-2 Molecular Formula: C5H3N5 Molecular Weight (g/mol): 133.11 MDL Number: MFCD00129982 InChI Key: MLOXIXGLIZLPDP-UHFFFAOYSA-N Synonym: 2-Amino-4,5-imidazoledicarbonitrile PubChem CID: 170446 IUPAC Name: 2-amino-1H-imidazole-4,5-dicarbonitrile SMILES: NC1=NC(C#N)=C(N1)C#N