accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 4096-21-3

IUPAC Name: 1-phenylpyrrolidine
Synonyms: 1-phenylpyrrolidine
Molecular Weight (g/mol): 147.22
Molecular Formula: C10H13N
InChi Key: VDQQJMHXZCMNMU-UHFFFAOYSA-N
SMILES: C1CCN(C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)