CAS RN 4097-88-5
IUPAC Name:
bis(2-aminoethyl)(methyl)amine
Synonyms:
bis(2-aminoethyl)(methyl)amine
Molecular Weight (g/mol):
117.2
Molecular Formula:
C5H15N3
InChi Key:
HYSQEYLBJYFNMH-UHFFFAOYSA-N
SMILES:
CN(CCN)CCN
1
–
1
of
1
results
1
2,2'-Diamino-N-methyldiethylamine 98.0+%, TCI America™
CAS: 4097-88-5 Molecular Formula: C5H15N3 Molecular Weight (g/mol): 117.20 MDL Number: MFCD00198039 InChI Key: HYSQEYLBJYFNMH-UHFFFAOYSA-N Synonym: N-(2-Aminoethyl)-N-methyl-1,2-ethanediamine, N,N-Bis(2-aminoethyl)methylamine PubChem CID: 430288 IUPAC Name: bis(2-aminoethyl)(methyl)amine SMILES: CN(CCN)CCN