accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3CH3SiSiCH3CH3CH3SiCH3CH3H3CSiCH3CH3H3C

CAS RN 4098-98-0

IUPAC Name: 1,1,1,3,3,3-hexamethyl-2,2-bis(trimethylsilyl)trisilane
Synonyms: 1,1,1,3,3,3-hexamethyl-2,2-bis(trimethylsilyl)trisilane
Molecular Weight (g/mol): 320.85
Molecular Formula: C12H36Si5
InChi Key: BOJSDHZZKKYWAS-UHFFFAOYSA-N
SMILES: C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (6)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)