accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40985-58-8

IUPAC Name: thieno[3,2-b]thiophene-2-carbonitrile
Synonyms: thieno[3,2-b]thiophene-2-carbonitrile
Molecular Weight (g/mol): 165.23
Molecular Formula: C7H3NS2
InChi Key: UPRBPGCXCNBTQH-UHFFFAOYSA-N
SMILES: N#CC1=CC2=C(S1)C=CS2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results