accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHIHO

CAS RN 41046-67-7

IUPAC Name: 2-iodobenzene-1,3-diol
Synonyms: 2-iodobenzene-1,3-diol
Molecular Weight (g/mol): 236.01
Molecular Formula: C6H5IO2
InChi Key: BNJXHRMYHDWZKL-UHFFFAOYSA-N
SMILES: OC1=CC=CC(O)=C1I
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)