accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 41051-15-4

IUPAC Name: methyl 4-methoxy-3-oxobutanoate
Synonyms: methyl 4-methoxy-3-oxobutanoate
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: QGBPKJFJAVDUNC-UHFFFAOYSA-N
SMILES: COCC(=O)CC(=O)OC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)