accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41052-75-9

IUPAC Name: hydrogen (2-chlorophenyl)hydrazine chloride
Synonyms: hydrogen (2-chlorophenyl)hydrazine chloride
Molecular Weight (g/mol): 179.04
Molecular Formula: C6H8Cl2N2
InChi Key: ADODRSVGNHNKAT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=CC=C1Cl
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)