accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH3CCH3CH3CH3CH3OHClOOO(E/Z)(E/Z)

CAS RN 41085-99-8

IUPAC Name: 2-[3-(3,3-dimethyl-1-octadecyl-2,3-dihydro-1H-indol-2-ylidene)prop-1-en-1-yl]-3,3-dimethyl-1-octadecyl-3H-indol-1-ium; perchloric acid
Synonyms: 2-[3-(3,3-dimethyl-1-octadecylindol-2-ylidene)prop-1-en-1-yl]-3,3-dimethyl-1-octadecylindol-1-ium; perchloric acid
Molecular Weight (g/mol): 934.89
Molecular Formula: C59H98ClN2O4
InChi Key: JVXZRNYCRFIEGV-UHFFFAOYSA-N
SMILES: O[Cl](=O)(=O)=O.CCCCCCCCCCCCCCCCCCN1C2=CC=CC=C2C(C)(C)C1=CC=CC1=[N+](CCCCCCCCCCCCCCCCCC)C2=CC=CC=C2C1(C)C
Quantity 
  • (1)
  • (1)
Color 
  • (1)
Label Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results