accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4110-50-3

IUPAC Name: 1-(ethylsulfanyl)propane
Synonyms: ethylpropylsulfide
Molecular Weight (g/mol): 104.21
Molecular Formula: C5H12S
InChi Key: ZDDDFDQTSXYYSE-UHFFFAOYSA-N
SMILES: CCCSCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results