accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN

CAS RN 41122-70-7

IUPAC Name: 4'-hexyl-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-hexyl-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 263.38
Molecular Formula: C19H21N
InChi Key: VADSDVGLFDVIMG-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results