accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41122-71-8

IUPAC Name: 4'-heptyl-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-heptyl-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 277.41
Molecular Formula: C20H23N
InChi Key: ZGOWXOZNUNZPAV-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results