accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CBr

CAS RN 4117-09-3

IUPAC Name: 7-bromohept-1-ene
Synonyms: 7-bromohept-1-ene
Molecular Weight (g/mol): 177.09
Molecular Formula: C7H13Br
InChi Key: GNYDYUQVALBGGZ-UHFFFAOYSA-N
SMILES: BrCCCCCC=C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)