accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41175-45-5

OON

CAS RN 41175-45-5

IUPAC Name: 3-oxa-16-azapentacyclo[10.7.1.0²,¹⁰.0⁵,⁹.0¹⁶,²⁰]icosa-1,5(9),10,12(20)-tetraen-4-one
Synonyms: 3-oxa-16-azapentacyclo[10.7.1.0²,¹⁰.0⁵,⁹.0¹⁶,²⁰]icosa-1,5(9),10,12(20)-tetraen-4-one
Molecular Weight (g/mol): 281.36
Molecular Formula: C18H19NO2
InChi Key: LLSRPENMALNOFW-UHFFFAOYSA-N
SMILES: O=C1OC2=C3CCCN4CCCC(C=C2C2=C1CCC2)=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results