accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOBr

CAS RN 41177-72-4

IUPAC Name: 5-bromo-2,3-dihydro-1-benzofuran-7-carboxylic acid
Synonyms: 5-bromo-2,3-dihydro-1-benzofuran-7-carboxylic acid
Molecular Weight (g/mol): 243.06
Molecular Formula: C9H7BrO3
InChi Key: LEBMKAXASFPSFA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=CC2=C1OCC2
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)