accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SPOOCH3OBrCl

CAS RN 41198-08-7

IUPAC Name: 4-bromo-2-chlorophenyl ethyl (propylsulfanyl)phosphonate
Synonyms: profenofos selecron caswell no. 266AA polycron epa pesticide chemical code 111401 curacron C11H15BrClO3PS
Molecular Weight (g/mol): 373.63
Molecular Formula: C11H15BrClO3PS
InChi Key: QYMMJNLHFKGANY-UHFFFAOYNA-N
SMILES: CCCSP(=O)(OCC)OC1=CC=C(Br)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results