accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3SiH3CH3CCH3CH3NBr

CAS RN 412048-44-3

IUPAC Name: 4-bromo-1-[tris(propan-2-yl)silyl]-1H-indole
Synonyms: 4-bromo-1-(triisopropylsilyl)indole
Molecular Weight (g/mol): 352.39
Molecular Formula: C17H26BrNSi
InChi Key: ZXHMUQVMGJYGAV-UHFFFAOYSA-N
SMILES: CC(C)[Si](C(C)C)(C(C)C)N1C=CC2=C(Br)C=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results