CAS RN 412307-34-7
IUPAC Name:
2-ethanethioamidoacetic acid
Synonyms:
ethanethioamidoacetic acid
Molecular Weight (g/mol):
133.17
Molecular Formula:
C4H7NO2S
InChi Key:
MOPCBCAABYPLOC-UHFFFAOYSA-N
SMILES:
CC(=S)NCC(O)=O
1
–
1
of
1
results
1
N-(1-Thioxoethyl)glycine 97.0+%, TCI America™
CAS: 412307-34-7 Molecular Formula: C4H7NO2S Molecular Weight (g/mol): 133.165 InChI Key: MOPCBCAABYPLOC-UHFFFAOYSA-N Synonym: 2-Ethanethioamidoacetic Acid PubChem CID: 14389889 IUPAC Name: 2-(ethanethioylamino)acetic acid SMILES: CC(=S)NCC(=O)O