accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOO

CAS RN 412357-50-7

IUPAC Name: benzyl N-(4-formylcyclohexyl)carbamate
Synonyms: benzyl N-(4-formylcyclohexyl)carbamate
Molecular Weight (g/mol): 261.32
Molecular Formula: C15H19NO3
InChi Key: SOIRQISQWQDGSN-UHFFFAOYSA-N
SMILES: O=CC1CCC(CC1)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)