accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO(E)

CAS RN 4124-88-3

IUPAC Name: (3E)-non-3-enoic acid
Synonyms: β-nonylenic acid 3-nonylenic acid
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: ZBPYTVBKHKUNHG-VOTSOKGWSA-N
SMILES: CCCCC\C=C\CC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results