accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOICl

CAS RN 41252-96-4

IUPAC Name: 2-chloro-1-iodo-4-nitrobenzene
Synonyms: 2-chloro-1-iodo-4-nitrobenzene
Molecular Weight (g/mol): 283.45
Molecular Formula: C6H3ClINO2
InChi Key: FQXFHRSYMORXKN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(I)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results