accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OON

CAS RN 41267-76-9

IUPAC Name: 6-methyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Synonyms: coumarin 102 coumarin 480
Molecular Weight (g/mol): 255.32
Molecular Formula: C16H17NO2
InChi Key: XHXMPURWMSJENN-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=C3CCCN4CCCC(C=C12)=C34
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)