accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNCl

CAS RN 41270-66-0

IUPAC Name: 5-chloro-2,3-diphenylpyrazine
Synonyms: 2,3-diphenyl-5-chloropyrazine
Molecular Weight (g/mol): 266.73
Molecular Formula: C16H11ClN2
InChi Key: VUGNCPVAXWZTOL-UHFFFAOYSA-N
SMILES: ClC1=CN=C(C2=CC=CC=C2)C(=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results